1-(4-Bromo-2,3-difluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

49046-500MG 49046-1G 49046-5G 49046-25G
77327-848EA 168.92 USD
77327-848 77346-262 77364-162 77386-552
1-(4-Bromo-2,3-difluorophenyl)ethanol ≥97%
1-(4-Bromo-2,3-difluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD28142179
CAS Number: 1823588-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR