3-Iodo-6-methoxy-2-methylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
49045-500MG
49045-1G
49045-5G
77337-622EA
431.57
USD
77337-622
77351-378
77375-642
3-Iodo-6-methoxy-2-methylpyridine ≥97%
3-Iodo-6-methoxy-2-methylpyridine
Precaución: for Research Use Only
Formula:
C₇H₈INO MW: 249.049 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16611481 CAS Number: 1092978-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...