(3-Bromo-2,6-dimethoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

49040-500MG 49040-1G 49040-5G 49040-25G
77327-846EA 279.13 USD
77327-846 77351-376 77364-160 77386-550
(3-Bromo-2,6-dimethoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(3-Bromo-2,6-dimethoxyphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₁₉BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34756538
CAS Number: 2755724-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR