(3-Bromo-2,6-dimethoxyphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

49039-500MG 49039-1G 49039-5G 49039-25G
77327-844EA 279.13 USD
77327-844 77351-374 77364-158 77386-548
(3-Bromo-2,6-dimethoxyphenyl)(thiomorpholino)methanone ≥97%
(3-Bromo-2,6-dimethoxyphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO₃S
MW: 346.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756537
CAS Number: 2755722-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR