(3-Bromo-2,6-dimethoxyphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

49038-500MG 49038-1G 49038-5G 49038-25G
77327-842EA 279.13 USD
77327-842 77351-372 77364-156 77386-546
(3-Bromo-2,6-dimethoxyphenyl)(morpholino)methanone ≥97%
(3-Bromo-2,6-dimethoxyphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO₄
MW: 330.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756536
CAS Number: 2755717-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR