(3-Bromo-2,6-dimethoxyphenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

49037-500MG 49037-1G 49037-5G 49037-25G
77327-840EA 279.13 USD
77327-840 77351-370 77364-154 77386-544
(3-Bromo-2,6-dimethoxyphenyl)(piperidin-1-yl)methanone ≥97%
(3-Bromo-2,6-dimethoxyphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD34756535
CAS Number: 2755723-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR