(3-Bromo-2,6-dimethoxyphenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

49036-500MG 49036-1G 49036-5G 49036-25G
77327-838EA 279.13 USD
77327-838 77351-368 77364-152 77386-542
(3-Bromo-2,6-dimethoxyphenyl)(pyrrolidin-1-yl)methanone ≥97%
(3-Bromo-2,6-dimethoxyphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD34756534
CAS Number: 2755721-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR