3-Bromo-N-cyclohexyl-2,6-dimethoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

49035-500MG 49035-1G 49035-5G
77327-836EA 310.03 USD
77327-836 77351-366 77375-640
3-Bromo-N-cyclohexyl-2,6-dimethoxybenzamide ≥97%
3-Bromo-N-cyclohexyl-2,6-dimethoxybenzamide

Precaución: for Research Use Only

Formula: C₁₅H₂₀BrNO₃
MW: 342.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756533
CAS Number: 2755716-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR