3-Bromo-N-isopropyl-2,6-dimethoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

49034-500MG 49034-1G 49034-25G 49034-5G
77327-834EA 301.79 USD
77327-834 77351-364 77386-540 77375-638
3-Bromo-N-isopropyl-2,6-dimethoxybenzamide ≥97%
3-Bromo-N-isopropyl-2,6-dimethoxybenzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD34756532
CAS Number: 2755722-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR