3-Bromo-N-cyclopropyl-2,6-dimethoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

49033-500MG 49033-1G 49033-5G
77327-832EA 310.03 USD
77327-832 77351-362 77375-636
3-Bromo-N-cyclopropyl-2,6-dimethoxybenzamide ≥97%
3-Bromo-N-cyclopropyl-2,6-dimethoxybenzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD34756531
CAS Number: 2755720-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR