Ethyl 4-ethoxy-2,6-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

48999-500MG 48999-1G 48999-5G 48999-25G
77327-818EA 230.72 USD
77327-818 77351-348 77364-140 77386-530
Ethyl 4-ethoxy-2,6-dimethylbenzoate ≥97%
Ethyl 4-ethoxy-2,6-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD17169501
CAS Number: 23811-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR