Ethyl 2-fluoro-3-(methylthio)benzoate ≥95%

Supplier: AOBChem USA

Warning

48978-500MG 48978-1G 48978-5G 48978-25G
77327-812EA 168.92 USD
77327-812 77346-260 77364-136 77386-526
Ethyl 2-fluoro-3-(methylthio)benzoate ≥95%
Ethyl 2-fluoro-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂S
Storage Temperature: Ambient
MDL Number: MFCD22489133
CAS Number: 1823046-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR