Methyl 3-ethoxy-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

48964-500MG 48964-1G 48964-5G 48964-25G
77327-804EA 140.08 USD
77327-804 77344-716 77364-126 77386-520
Methyl 3-ethoxy-4-methylbenzoate ≥97%
Methyl 3-ethoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34529992
CAS Number: 87808-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR