(3-Ethoxy-4-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48963-500MG 48963-1G 48963-5G 48963-25G
77327-802EA 280.16 USD
77327-802 77351-342 77364-124 77386-518
(3-Ethoxy-4-methylphenyl)methanol ≥97%
(3-Ethoxy-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD30078686
CAS Number: 1823878-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR