4-Fluoro-3-isopropoxyphenol ≥95%

Supplier: AOBChem USA

Warning

48960-500MG 48960-1G 48960-5G
77338-596EA 503.67 USD
77338-596 77351-338 77375-622
4-Fluoro-3-isopropoxyphenol ≥95%
4-Fluoro-3-isopropoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD16997325
CAS Number: 1243390-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR