Ethyl 4-fluoro-3-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

48959-500MG 48959-1G 48959-5G 48959-25G
77327-796EA 140.08 USD
77327-796 77344-714 77364-118 77386-512
Ethyl 4-fluoro-3-isopropoxybenzoate ≥97%
Ethyl 4-fluoro-3-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃
MW: 226.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23114968
CAS Number: 1394960-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR