3-(Benzyloxy)-5-bromo-2-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

48958-500MG 48958-1G 48958-5G
77337-878EA 450.11 USD
77337-878 77351-336 77375-620
3-(Benzyloxy)-5-bromo-2-fluorophenol ≥97%
3-(Benzyloxy)-5-bromo-2-fluorophenol

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrFO₂
MW: 297.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529991
CAS Number: 2734779-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR