Ethyl 3-(benzyloxy)-5-bromo-2-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

48955-500MG 48955-1G 48955-25G 48955-5G
77327-794EA 302.82 USD
77327-794 77351-332 77386-510 77375-616
Ethyl 3-(benzyloxy)-5-bromo-2-fluorobenzoate ≥95%
Ethyl 3-(benzyloxy)-5-bromo-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₄BrFO₃
MW: 353.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529990
CAS Number: 2734779-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR