Methyl 3-(benzyloxy)-5-bromo-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

48954-500MG 48954-1G 48954-5G 48954-25G
77327-792EA 278.1 USD
77327-792 77351-330 77364-116 77386-508
Methyl 3-(benzyloxy)-5-bromo-2-fluorobenzoate ≥97%
Methyl 3-(benzyloxy)-5-bromo-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₂BrFO₃
MW: 339.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529989
CAS Number: 2734775-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR