3-(Benzyloxy)-5-bromo-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48953-500MG 48953-1G 48953-5G 48953-25G
77327-790EA 233.81 USD
77327-790 77351-328 77364-114 77386-506
3-(Benzyloxy)-5-bromo-2-fluorobenzoic acid ≥97%
3-(Benzyloxy)-5-bromo-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrFO₃
MW: 325.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902791
CAS Number: 2586127-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR