(3-(Benzyloxy)-5-bromo-2-fluorophenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
48948-500MG
48948-1G
48948-5G
48948-25G
77327-786EA
124.63
USD
77327-786
77344-074
77364-110
77375-610
(3-(Benzyloxy)-5-bromo-2-fluorophenyl)(methyl)sulfane ≥97%
(3-(Benzyloxy)-5-bromo-2-fluorophenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₁₄H₁₂BrFOS MW: 327.212 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD33403923 CAS Number: 2624416-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...