5-Bromo-2-fluoro-1-iodo-3-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

48946-500MG 48946-1G 48946-5G 48946-25G
77327-784EA 186.43 USD
77327-784 77351-324 77364-108 77386-504
5-Bromo-2-fluoro-1-iodo-3-isopropoxybenzene ≥97%
5-Bromo-2-fluoro-1-iodo-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrFIO
MW: 358.974 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902789
CAS Number: 2586126-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR