Ethyl 5-bromo-2-fluoro-3-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

48942-500MG 48942-1G 48942-5G 48942-25G
77327-780EA 121.54 USD
77327-780 77343-916 77364-104 77375-606
Ethyl 5-bromo-2-fluoro-3-isopropoxybenzoate ≥97%
Ethyl 5-bromo-2-fluoro-3-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrFO₃
MW: 305.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403921
CAS Number: 2624417-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR