Methyl 5-bromo-2-fluoro-3-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

48941-500MG 48941-1G 48941-5G 48941-25G
77327-778EA 115.36 USD
77327-778 77343-396 77364-102 77375-604
Methyl 5-bromo-2-fluoro-3-isopropoxybenzoate ≥97%
Methyl 5-bromo-2-fluoro-3-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrFO₃
MW: 291.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403920
CAS Number: 2624417-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR