(5-Bromo-2-fluoro-3-isopropoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48939-500MG 48939-1G 48939-5G 48939-25G
77327-774EA 225.57 USD
77327-774 77351-322 77364-098 77386-502
(5-Bromo-2-fluoro-3-isopropoxyphenyl)methanol ≥97%
(5-Bromo-2-fluoro-3-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
MW: 263.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756519
CAS Number: 2764729-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR