2-Chloro-5-methoxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48907-500MG 48907-1G 48907-5G 48907-25G
77327-766EA 281.19 USD
77327-766 77351-318 77364-090 77386-494
2-Chloro-5-methoxy-3-methylbenzoic acid ≥97%
2-Chloro-5-methoxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756504
CAS Number: 1785263-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR