1-Bromo-2-chloro-5-methoxy-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

48904-500MG 48904-1G 48904-5G 48904-25G
77327-764EA 157.59 USD
77327-764 77345-694 77364-088 77386-492
1-Bromo-2-chloro-5-methoxy-3-methylbenzene ≥95%
1-Bromo-2-chloro-5-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756501
CAS Number: 1785028-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR