1-Bromo-2-iodo-5-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

48902-500MG 48902-1G 48902-5G 48902-25G
77327-762EA 207.03 USD
77327-762 77351-316 77364-086 77386-490
1-Bromo-2-iodo-5-methoxy-3-methylbenzene ≥97%
1-Bromo-2-iodo-5-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrIO
Storage Temperature: Ambient
MDL Number: MFCD34756499
CAS Number: 2383587-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR