(2-Fluoro-5-iodo-4-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48895-500MG 48895-1G 48895-5G 48895-25G
77327-760EA 181.28 USD
77327-760 77346-884 77364-084 77386-488
(2-Fluoro-5-iodo-4-methylphenyl)methanol ≥97%
(2-Fluoro-5-iodo-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈FIO
MW: 266.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31543083
CAS Number: 2386997-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR