(2-Fluoro-5-iodo-3-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48886-500MG 48886-1G 48886-5G 48886-25G
77327-754EA 118.45 USD
77327-754 77343-556 77364-078 77375-596
(2-Fluoro-5-iodo-3-methylphenyl)methanol ≥97%
(2-Fluoro-5-iodo-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈FIO
MW: 266.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756495
CAS Number: 2385917-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR