Ethyl 2-fluoro-5-iodo-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

48884-500MG 48884-1G 48884-5G 48884-25G
77327-752EA 139.05 USD
77327-752 77344-666 77364-076 77386-484
Ethyl 2-fluoro-5-iodo-3-methylbenzoate ≥97%
Ethyl 2-fluoro-5-iodo-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀FIO₂
Storage Temperature: Ambient
MDL Number: MFCD24115509
CAS Number: 1508552-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR