Methyl 2-fluoro-5-iodo-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

48883-500MG 48883-1G 48883-5G 48883-25G
77327-750EA 136.99 USD
77327-750 77344-592 77364-074 77386-482
Methyl 2-fluoro-5-iodo-3-methylbenzoate ≥97%
Methyl 2-fluoro-5-iodo-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈FIO₂
MW: 294.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24115505
CAS Number: 1542900-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR