(3-Bromo-2-fluoro-5-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48878-500MG 48878-1G 48878-5G 48878-25G
77327-746EA 129.78 USD
77327-746 77344-318 77364-068 77375-590
(3-Bromo-2-fluoro-5-iodophenyl)methanol ≥97%
(3-Bromo-2-fluoro-5-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrFIO
MW: 330.921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756493
CAS Number: 2090447-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR