(2,5-dimethoxy-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

48868-500MG 48868-1G 48868-5G 48868-25G
77327-740EA 225.57 USD
77327-740 77351-310 77364-062 77386-476
(2,5-dimethoxy-4-methylphenyl)(methyl)sulfane ≥97%
(2,5-dimethoxy-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD32902778
CAS Number: 500364-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR