3,6-Dichloro-2-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48863-500MG 48863-1G 48863-5G 48863-25G
77327-738EA 201.88 USD
77327-738 77351-308 77364-060 77386-474
3,6-Dichloro-2-hydroxybenzaldehyde ≥97%
3,6-Dichloro-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01666394
CAS Number: 27164-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR