2,4,6-trifluoro-3-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

48856-500MG 48856-1G 48856-5G 48856-25G
77327-736EA 206 USD
77327-736 77351-306 77364-058 77386-472
2,4,6-trifluoro-3-methoxyphenol ≥95%
2,4,6-trifluoro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₅F₃O₂
MW: 178.109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28800132
CAS Number: 1804096-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR