Methyl 6-chloro-2-fluoro-3-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

48830-500MG 48830-1G 48830-5G 48830-25G
77327-732EA 90.64 USD
77327-732 77342-388 77364-054 77375-582
Methyl 6-chloro-2-fluoro-3-iodobenzoate ≥97%
Methyl 6-chloro-2-fluoro-3-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₅ClFIO₂
Storage Temperature: Ambient
MDL Number: MFCD33403919
CAS Number: 2091029-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR