Ethyl 6-chloro-2-fluoro-3-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

48829-500MG 48829-1G 48829-5G 48829-25G
77327-730EA 150.38 USD
77327-730 77345-344 77364-052 77386-468
Ethyl 6-chloro-2-fluoro-3-iodobenzoate ≥97%
Ethyl 6-chloro-2-fluoro-3-iodobenzoate

Precaución: for Research Use Only

Formula: C₉H₇ClFIO₂
Storage Temperature: Ambient
MDL Number: MFCD34564129
CAS Number: 2384435-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR