Methyl 3-bromo-5-fluoro-2-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

48826-500MG 48826-1G 48826-5G
77338-628EA 504.7 USD
77338-628 77351-300 77375-580
Methyl 3-bromo-5-fluoro-2-isopropoxybenzoate ≥97%
Methyl 3-bromo-5-fluoro-2-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrFO₃
MW: 291.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178551
CAS Number: 2119355-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR