Methyl 4,5-difluoro-2-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

48823-500MG 48823-1G 48823-25G 48823-5G
77327-728EA 298.7 USD
77327-728 77351-298 77386-466 77375-578
Methyl 4,5-difluoro-2-isopropoxybenzoate ≥97%
Methyl 4,5-difluoro-2-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
MW: 230.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23114998
CAS Number: 1394955-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR