Ethyl 4,5-difluoro-2-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

48822-500MG 48822-1G 48822-5G
77327-726EA 318.27 USD
77327-726 77351-296 77375-576
Ethyl 4,5-difluoro-2-isopropoxybenzoate ≥97%
Ethyl 4,5-difluoro-2-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂O₃
MW: 244.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403917
CAS Number: 2107512-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR