Ethyl 3-bromo-2-fluoro-6-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

48820-500MG 48820-1G 48820-5G 48820-25G
77327-724EA 189.52 USD
77327-724 77351-294 77364-050 77386-464
Ethyl 3-bromo-2-fluoro-6-(trifluoromethyl)benzoate ≥97%
Ethyl 3-bromo-2-fluoro-6-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₇BrF₄O₂
MW: 315.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902773
CAS Number: 2586126-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR