Methyl 3-bromo-2-fluoro-6-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

48819-500MG 48819-1G 48819-5G 48819-25G
77327-722EA 100.94 USD
77327-722 77342-920 77364-048 77375-574
Methyl 3-bromo-2-fluoro-6-(trifluoromethyl)benzoate ≥97%
Methyl 3-bromo-2-fluoro-6-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₅BrF₄O₂
MW: 301.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902772
CAS Number: 2090828-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR