(3-Bromo-2-fluoro-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
48816-500MG
48816-1G
48816-5G
48816-25G
77327-720EA
97.85
USD
77327-720
77342-668
77364-046
77375-572
(3-Bromo-2-fluoro-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(3-Bromo-2-fluoro-6-(trifluoromethyl)phenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₈H₅BrF₄S Storage Temperature: Ambient |
MDL Number:
MFCD32902771 CAS Number: 2166849-71-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...