Ethyl 4-(benzyloxy)-3-chloro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

48814-500MG 48814-1G 48814-25G 48814-5G
77327-718EA 297.67 USD
77327-718 77351-292 77386-462 77375-570
Ethyl 4-(benzyloxy)-3-chloro-2-methylbenzoate ≥97%
Ethyl 4-(benzyloxy)-3-chloro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD00598351
CAS Number: 2635937-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR