4-(Benzyloxy)-3-chloro-2-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

48808-500MG 48808-1G 48808-5G
77339-352EA 581.95 USD
77339-352 77351-284 77375-562
4-(Benzyloxy)-3-chloro-2-(methylthio)benzaldehyde ≥95%
4-(Benzyloxy)-3-chloro-2-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₂S
MW: 292.781 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902769
CAS Number: 2586126-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR