(3-Fluoro-4-methoxy-2-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48801-500MG 48801-1G 48801-5G 48801-25G
77327-712EA 281.19 USD
77327-712 77351-282 77364-042 77386-458
(3-Fluoro-4-methoxy-2-(methylthio)phenyl)methanol ≥97%
(3-Fluoro-4-methoxy-2-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂S
Storage Temperature: Ambient
MDL Number: MFCD33403912
CAS Number: 2090969-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR