3-Fluoro-4-methoxy-2-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

48800-500MG 48800-1G 48800-5G 48800-25G
77327-710EA 177.16 USD
77327-710 77346-592 77364-040 77386-456
3-Fluoro-4-methoxy-2-(methylthio)benzaldehyde ≥95%
3-Fluoro-4-methoxy-2-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902767
CAS Number: 2586126-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR