Ethyl 3-fluoro-4-methoxy-2-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

48799-500MG 48799-1G 48799-5G 48799-25G
77327-708EA 280.16 USD
77327-708 77351-280 77364-038 77386-454
Ethyl 3-fluoro-4-methoxy-2-(methylthio)benzoate ≥97%
Ethyl 3-fluoro-4-methoxy-2-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃S
MW: 244.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403911
CAS Number: 2635937-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR