Methyl 3-fluoro-4-methoxy-2-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

48798-500MG 48798-1G 48798-5G 48798-25G
77327-706EA 280.16 USD
77327-706 77351-278 77364-036 77386-452
Methyl 3-fluoro-4-methoxy-2-(methylthio)benzoate ≥97%
Methyl 3-fluoro-4-methoxy-2-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃S
MW: 230.256 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403910
CAS Number: 2635937-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR